Dioctyl sulfide ≥97.0%

Supplier: TCI America
Danger

Synonyms: Dioctylsulfane, Dioctylsulphane

O0108-10G O0108-100G
TCO0108-010GEA 30.02 CAD
TCO0108-010G TCO0108-100G
Dioctyl sulfide ≥97.0%
Dioctyl sulphide

CAS Number: 2690-08-6
MDL Number: MFCD00009566
Molecular Formula: C16H34S
Molecular Weight: 258.51
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 309
Flash Point (°C): 160
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: Di-n-octyl Sulfide

Formula: C₁₆H₃₄S
MW: 258.51 g/mol
Boiling Pt: 309 °C
Density: 0.85 (20 °C)
Flash Pt: 160 °C
MDL Number: MFCD00009566
CAS Number: 2690-08-6
EINECS: 220-259-5
UN: 3334
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c63ea24b35cf3af841e71bbfa2cc5d4bfbc4f97c&vid=c63ea24b35cf3af841e71bbfa2cc5d4bfbc4f97c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR