Oxamide ≥98.0% (by total nitrogen basis)
Supplier: TCI America
O0086-25G
O0086-500G
O0086-100G
TCO0086-025GEA
26.86
CAD
TCO0086-025G
TCO0086-500G
TCO0086-100G
Oxamide ≥98.0% (by total nitrogen basis)
Oxamide
CAS Number: 471-46-5
MDL Number: MFCD00008007
Molecular Formula: C2H4N2O2
Molecular Weight: 88.07
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Formula:
C₂H₄N₂O₂ MW: 88.07 g/mol |
MDL Number:
MFCD00008007 CAS Number: 471-46-5 EINECS: 207-442-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...