Oxamic acid ≥97.0% (by total nitrogen and titration analysis)

Supplier: TCI America

Synonyms: 2-Amino-2-oxoacetic acid

O0084-5G O0084-25G
TCO0084-5GEA 50.56 CAD
TCO0084-5G TCO0084-025G
Oxamic acid ≥97.0% (by total nitrogen and titration analysis)
Oxamic acid

CAS Number: 471-47-6
MDL Number: MFCD00008006
Molecular Formula: C2H3NO3
Molecular Weight: 89.05
Purity/Analysis Method: >97.0% (T,N)
Form: Crystal
Melting point (°C): 217


Other Product Names / Synonyms: Oxamidic Acid

Formula: C₂H₃NO₃
MW: 89.05 g/mol
Melting Pt: 217 °C
MDL Number: MFCD00008006
CAS Number: 471-47-6
EINECS: 207-443-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da17d4f6da9746d4f172e349a94fea1d59ca7b34&vid=da17d4f6da9746d4f172e349a94fea1d59ca7b34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR