Dimethyl oxalate ≥99.0%

Supplier: TCI America
Warning

Synonyms: Oxalic acid dimethyl ester

O0080-500G O0080-25G
TCO0080-500GEA 104.28 CAD
TCO0080-500G TCO0080-25G
Dimethyl oxalate ≥99.0%
Dimethyl oxalate

CAS Number: 553-90-2
MDL Number: MFCD00008442
Molecular Formula: C4H6O4
Molecular Weight: 118.09
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 164
Melting point (°C): 54
Flash Point (°C): 67


Other Product Names / Synonyms: Oxalic Acid Dimethyl Ester

Formula: C₄H₆O₄
MW: 118.09 g/mol
Boiling Pt: 163
Melting Pt: 54
Density: 1.422 g/cm³ (15 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00008442
CAS Number: 553-90-2
EINECS: 209-053-6
UN: 3261
ADR: 8 III
Merck Index: 12,06181
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd9626343cf0139c93b3e184c1e18509d2c1d717&vid=bd9626343cf0139c93b3e184c1e18509d2c1d717&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR