Orotic acid monohydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

O0065-500G O0065-25G O0065-100G
TCO0065-500GEA 271.76 CAD
TCO0065-500G TCO0065-025G TCO0065-100G
Orotic acid monohydrate ≥98.0% (by HPLC, titration analysis)
Orotic acid monohydrate

CAS Number: 50887-69-9
MDL Number: MFCD00006027
Molecular Formula: C5H4N2O4
Molecular Weight: 156.10
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 6-Carboxyuracil, Uracil-6-carboxylic Acid, Vitamin B13

Formula: C₅H₄N₂O₄·1H₂O
MDL Number: MFCD00149398
CAS Number: 50887-69-9
EINECS: 200-619-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b05f1710918dec04715f78da776bc1dbb2d4441&vid=1b05f1710918dec04715f78da776bc1dbb2d4441&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR