L(+)-Ornithine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: H-Orn-OH Hydrochloride, H-Orn-OH·HCl

O0064-250G O0064-25G
TCO0064-250GEA 287.56 CAD
TCO0064-250G TCO0064-025G
L(+)-Ornithine hydrochloride ≥98.0% (by HPLC, titration analysis)
L(+)-Ornithine hydrochloride

CAS Number: 3184-13-2
MDL Number: MFCD00064562
Molecular Formula: C5H12N2O2
Molecular Weight: 168.62
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 24 deg (C=4, 6mol/L HCl)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids, Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-2,5-Diaminopentanoic Acid Monohydrochloride, (S)-2,5-Diaminovaleric Acid Monohydrochloride, H-Orn-OH.HCl

Formula: C₅H₁₃ClN₂O₂
MW: 168.62 g/mol
MDL Number: MFCD00064562
CAS Number: 3184-13-2
EINECS: 221-678-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR