Oleyl alcohol 60.0% (by GC)

Supplier: TCI America
Warning

Synonyms: cis-Octadec-9-en-1-olcis-9-Octadecen-1-ol

O0058-25ML O0058-500ML
TCO0058-025MLEA 34.37 CAD
TCO0058-025ML TCO0058-500ML
Oleyl alcohol 60.0% (by GC)
Oleyl alcohol

CAS Number: 143-28-2
MDL Number: MFCD00002993
Molecular Formula: C18H36O
Molecular Weight: 268.49
Purity/Analysis Method: >60.0% (GC)
Form: Clear Liquid
Boiling point (°C): 207
Flash Point (°C): 189
Specific Gravity (20/20): 0.85

Formula: C₁₈H₃₆O
MW: 268.48 g/mol
Boiling Pt: 207 °C
Melting Pt: 13…19 °C
Density: 0.85 (20 °C)
Flash Pt: 189 °C
MDL Number: MFCD00002993
CAS Number: 143-28-2
Merck Index: 12,06968
https://ca.vwr.com/stibo/web/std.lang.all/03/69/28990369.jpg

Order Now

Specification Test Results

Purity/Analysis Method 60.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR