Methyl oleate 60.0% (by GC)

Supplier: TCI America

Synonyms: cis-Oleic Acid Methyl Ester, Oleic acid methyl ester

O0055-25ML O0055-500ML
TCO0055-025MLEA 31.6 CAD
TCO0055-025ML TCO0055-500ML
Methyl oleate 60.0% (by GC)
Methyl oleate

Molecular Weight: 296.50
Purity/Analysis Method: >60.0% (GC)
Form: Clear Liquid
Boiling point (°C): 161
Melting point (°C): -20
Flash Point (°C): 163
Specific Gravity (20/20): 0.88


Product Categories: Materials Chemistry, Resin Additives, Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Oleic Acid Methyl Ester

Formula: C₁₉H₃₆O₂
MW: 296.49 g/mol
Boiling Pt: 161 °C
Melting Pt: –20 °C
Density: 0.88 (20 °C)
Flash Pt: 163 °C
Storage Temperature: Ambient
MDL Number: MFCD00009578
CAS Number: 112-62-9
EINECS: 203-992-5
Merck Index: 12,06965
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e75b31f66687a40720249c215794d741e16d53bb&vid=e75b31f66687a40720249c215794d741e16d53bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 60.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR