Ethyl oleate ≥70.0%

Supplier: TCI America

O0054-25ML O0054-500ML
TCO0054-025MLEA 43.42 CAD
TCO0054-025ML TCO0054-500ML
Ethyl oleate ≥70.0%
Ethyl oleate

CAS Number: 111-62-6
MDL Number: MFCD00009579
Molecular Formula: C20H38O2
Molecular Weight: 310.52
Purity/Analysis Method: >70.0% (GC)
Form: Clear Liquid
Boiling point (°C): 209
Flash Point (°C): 181
Specific Gravity (20/20): 0.87


Product Categories: Materials Chemistry, Resin Additives, Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Oleic Acid Ethyl Ester

Formula: C₂₀H₃₈O₂
Boiling Pt: 209 °C
Melting Pt: –32 °C
Density: 0.87 (20 °C)
Flash Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD00009579
CAS Number: 111-62-6
EINECS: 203-889-5
Merck Index: 12,06965
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3719a336b93ed28720acf80973f3d2f4c954fdd7&vid=3719a336b93ed28720acf80973f3d2f4c954fdd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 70.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR