Octylbenzene ≥98.0%

Supplier: TCI America

Synonyms: 1-Phenyloctane

O0046-25ML O0046-500ML O0046-100ML
TCO0046-025MLEA 146.94 CAD
TCO0046-025ML TCO0046-500ML TCO0046-100ML
Octylbenzene ≥98.0%
Octylbenzene

CAS Number: 2189-60-8
MDL Number: MFCD00009564
Molecular Formula: C14H22
Molecular Weight: 190.33
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 264
Flash Point (°C): 107
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: 1-Phenyloctane

Formula: C₁₄H₂₂
Boiling Pt: 264 °C
Melting Pt: –3 °C
Density: 0.86 (20 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00009564
CAS Number: 2189-60-8
EINECS: 218-582-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bcafb144c86228f4b51240d279548ced56970b4&vid=b7a76249c68ec6be1a5e7e18efb1fed3e8178182&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR