1-Octanol ≥99.0%

Supplier: TCI America
Warning

Synonyms: n-Octanol

O0036-25ML O0036-500ML
TCO0036-25MLEA 37.19 CAD
TCO0036-25ML TCO0036-500ML
1-Octanol ≥99.0%
1-Octanol

CAS Number: 111-87-5
MDL Number: MFCD00002988
Molecular Formula: C8H18O
Molecular Weight: 130.23
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 194
Melting point (°C): -16
Flash Point (°C): 83
Specific Gravity (20/20): 0.83


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: n-Octyl Alcohol

Formula: H₃C(CH₂)₆CH₂OH
MW: 130.23 g/mol
Boiling Pt: 194 °C
Melting Pt: –16 °C
Density: 0.83 (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00002988
CAS Number: 111-87-5
EINECS: 203-917-6
Merck Index: 13,06784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9584627f8f6f6ecf55152882b8588f2e0243e819&vid=c35011278d1014420092a0c07be1ccd7b82d3478&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR