Ethyl octanoate ≥98.0%

Supplier: TCI America

Synonyms: Caprylic acid ethyl ester, Octanoic acid ethyl ester, Ethyl caprylate

O0030-500ML O0030-25ML
TCO0030-500MLEA 116.92 CAD
TCO0030-500ML TCO0030-025ML
Ethyl octanoate ≥98.0%
Ethyl octanoate

CAS Number: 106-32-1
MDL Number: MFCD00009552
Molecular Formula: C10H20O2
Molecular Weight: 172.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Melting point (°C): -43
Flash Point (°C): 92
Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: n-Octanoic Acid Ethyl Ester

Formula: C₁₀H₂₀O₂
Boiling Pt: 208 °C
Melting Pt: –43 °C
Density: 0.87 (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00009552
CAS Number: 106-32-1
EINECS: 203-385-5
Merck Index: 13,03813
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d912c3816db80d1a2fc5eeb17e2ca9c2999b1d9&vid=8d912c3816db80d1a2fc5eeb17e2ca9c2999b1d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR