1,8-Octanediol ≥99.0%
Supplier: TCI America
O0024-25G
O0024-500G
O0024-100G
TCO0024-025GEA
85.95
CAD
TCO0024-025G
TCO0024-500G
TCO0024-100G
1,8-Octanediol ≥99.0%
1,8-Octanediol
CAS Number: 629-41-4
MDL Number: MFCD00002989
Molecular Formula: C8H18O2
Molecular Weight: 146.23
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 166
Melting point (°C): 62
Flash Point (°C): 148
Other Product Names / Synonyms: 1,8-Dihydroxyoctane, Octamethylene Glycol
Formula:
C₈H₁₈O₂ Boiling Pt: 166 °C Melting Pt: 62 °C Density: 0.941 g/cm³ (20 °C) Flash Pt: 148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002989 CAS Number: 629-41-4 EINECS: 211-090-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...