Elaidic acid ≥97.0%
Supplier: TCI America
Synonyms:
(E)-Octadec-9-enoic acid
O0010-25G
O0010-5G
TCO0010-025GEA
721.54
CAD
TCO0010-025G
TCO0010-5G
Elaidic acid ≥97.0%
Elaidic acid
CAS Number: 112-79-8
MDL Number: MFCD00063954
Molecular Formula: C18H34O2
Molecular Weight: 282.47
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Boiling point (°C): 288
Melting point (°C): 44
Flash Point (°C): 88
Storage Temperature: 0-10°C
Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: trans-9-Octadecenoic Acid
Formula:
C₁₈H₃₄O₂ Boiling Pt: 288 °C Melting Pt: 44 °C Flash Pt: 88 °C |
MDL Number:
MFCD00063954 CAS Number: 112-79-8 EINECS: 204-006-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...