1-Octadecanol ≥98.0%

Supplier: TCI America

O0006-25G O0006-500G
TCO0006-025GEA 41.32 CAD
TCO0006-025G TCO0006-500G
1-Octadecanol ≥98.0%
1-Octadecanol

CAS Number: 112-92-5
MDL Number: MFCD00002823
Molecular Formula: C18H38O
Molecular Weight: 270.50
Purity/Analysis Method: >98.0% (GC)
Form: Pellet
Boiling point (°C): 210
Melting point (°C): 59
Flash Point (°C): 174


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Stearyl Alcohol, Octadecyl Alcohol

Formula: H₃C(CH₂)₁₆CH₂OH
Boiling Pt: 210 °C
Melting Pt: 59 °C
Density: 0.805…0.815 g/cm³ (60 °C)
Flash Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00002823
CAS Number: 112-92-5
EINECS: 204-017-6
Merck Index: 12,08960
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae3c0d98d76313e7abaaca48e836ab650af22048&vid=b5affc45efce06be9d2f2859a74da5ea08c251e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR