1-Octadecanethiol ≥97.0%

Supplier: TCI America

Synonyms: n-Octadecyl mercaptan, n-Octadecanethiol, Stearyl mercaptane, 1-Mercaptooctadecane, Octadecane-1-thiol

O0005-25G O0005-400G
TCO0005-025GEA 44.56 CAD
TCO0005-025G TCO0005-400G
1-Octadecanethiol ≥97.0%
1-Octadecanethiol

CAS Number: 2885-00-9
MDL Number: MFCD00004886
Molecular Formula: C18H38S
Molecular Weight: 286.56
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Flash Point (°C): 185
Freezing point (°C): 27


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Stearyl Mercaptan, Octadecyl Mercaptan

Formula: C₁₈H₃₈S
MW: 286.57 g/mol
Boiling Pt: 204
Melting Pt: 28…31 °C
Density: 0.8475 g/cm³ (20 °C)
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00004886
CAS Number: 2885-00-9
EINECS: 220-744-1
UN: 3335
ADR: 9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f04684306df4b8f4e8272da6cb37eaf24ead5e0e&vid=0755e1ffa0e49db104852ccd7088ba407d66f62d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR