n-Octadecane ≥98.0%
Supplier: TCI America
O0003-25G
O0003-250G
TCO0003-025GEA
42.96
CAD
TCO0003-025G
TCO0003-250G
n-Octadecane ≥98.0%
n-Octadecane
CAS Number: 593-45-3
MDL Number: MFCD00009007
Molecular Formula: C18H38
Molecular Weight: 254.50
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 166
Freezing point (°C): 28
Formula:
H₃C(CH₂)₁₆CH₃ Boiling Pt: 316.1 °C (1013 hPa) Melting Pt: 26…29 °C Density: 0.7765 g/cm³ (20 °C) Flash Pt: 166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009007 CAS Number: 593-45-3 EINECS: 209-790-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...