Ethyl nitroacetate ≥95.0%

Supplier: TCI America

Synonyms: Nitroethansäureethylester, Nitroacetic ethyl ester

N0657-25G N0657-5G
TCN0657-025GEA 260.7 CAD
TCN0657-025G TCN0657-005G
Ethyl nitroacetate ≥95.0%
Ethyl nitroacetate

CAS Number: 626-35-7
MDL Number: MFCD00007403
Molecular Formula: C4H7NO4
Molecular Weight: 133.10
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 107
Flash Point (°C): 92
Specific Gravity (20/20): 1.20


Other Product Names / Synonyms: Nitroacetic Acid Ethyl Ester

Formula: C₄H₇NO₄
MW: 133.1 g/mol
Boiling Pt: 107 °C
Density: 1.20 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00007403
CAS Number: 626-35-7
EINECS: 210-944-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74ffe933382ea8afbd9182888760fbbba54fdb8b&vid=74ffe933382ea8afbd9182888760fbbba54fdb8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR