Methyl nitroacetate ≥97.0%

Supplier: TCI America

Synonyms: Nitroacetic methyl ester, Methyl 2-nitroacetate

N0656-5G N0656-25G
TCN0656-005GEA 142.2 CAD
TCN0656-005G TCN0656-25G
Methyl nitroacetate ≥97.0%
Methyl nitroacetate

CAS Number: 2483-57-0
MDL Number: MFCD00075480
Molecular Formula: C3H5NO4
Molecular Weight: 119.08
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 95
Specific Gravity (20/20): 1.29


Other Product Names / Synonyms: Nitroacetic Acid Methyl Ester

Formula: C₃H₅NO₄
Boiling Pt: 100 °C
Density: 1.29 (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00075480
CAS Number: 2483-57-0
EINECS: 219-622-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eff02daee5c92c2679d1c5c5b31ffcb945efc4e&vid=6eff02daee5c92c2679d1c5c5b31ffcb945efc4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR