2-Naphthyl boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 2-Naphthaleneboronic acid, Naphthalene-2-boronic acid, 2-Naphthyl boric acid

N0649-5G N0649-1G N0649-25G
TCN0649-5GEA 95.91 CAD
TCN0649-5G TCN0649-001G TCN0649-25G
2-Naphthyl boronic acid (contains varying amounts of Anhydride)
2-Naphthyl boronic acid

CAS Number: 32316-92-0
MDL Number: MFCD00236051
Molecular Formula: C10H9BO2
Molecular Weight: 171.99
Form: Crystal
Color: White
Melting point (°C): 268


Other Product Names / Synonyms: 2-Naphthylboronic Acid

Formula: C₁₀H₉BO₂
MW: 171.99 g/mol
Melting Pt: 268 °C
MDL Number: MFCD00236051
CAS Number: 32316-92-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75c5084066fa850315e8522fec3e4275e308958d&vid=75c5084066fa850315e8522fec3e4275e308958d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR