5-Nitroanthranilic acid ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
2-Amino-5-nitrobenzoic acid
N0644-25G
N0644-5G
TCN0644-025GEA
137.46
CAD
TCN0644-025G
TCN0644-005G
5-Nitroanthranilic acid ≥98.0%
5-Nitroanthranilic acid
CAS Number: 616-79-5
MDL Number: MFCD00017039
Molecular Formula: C7H6N2O4
Molecular Weight: 182.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 2-Amino-5-nitrobenzoic Acid
Formula:
C₇H₆N₂O₄ Melting Pt: 278 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017039 CAS Number: 616-79-5 EINECS: 210-493-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...