Monomethyl-5-nitroisophthalate ≥98.0%

Supplier: TCI America

Synonyms: 5-Nitroisophthalic acid monomethyl ester, Methyl 5-nitroisophthalate

N0641-500G N0641-25G
TCN0641-500GEA 537.2 CAD
TCN0641-500G TCN0641-025G
Monomethyl-5-nitroisophthalate ≥98.0%
Monomethyl 5-nitroisophthalate

CAS Number: 1955-46-0
MDL Number: MFCD00009793
Molecular Formula: C9H7NO6
Molecular Weight: 225.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 182
Flash Point (°C): 120


Other Product Names / Synonyms: Methyl Hydrogen 5-Nitroisophthalate, 5-Nitroisophthalic Acid Monomethyl Ester

Formula: C₉H₇NO₆
Melting Pt: 182 °C
Flash Pt: 120 °C
MDL Number: MFCD00009793
CAS Number: 1955-46-0
EINECS: 217-793-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50fadb7b12ee304975e947bebb722c35a8079783&vid=50fadb7b12ee304975e947bebb722c35a8079783&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR