1-Naphthyl boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

N0630-1G N0630-25G N0630-5G
TCN0630-001GEA 33.18 CAD
TCN0630-001G TCN0630-25G TCN0630-005G
1-Naphthyl boronic acid (contains varying amounts of Anhydride)
1-Naphthyl boronic acid

CAS Number: 13922-41-3
MDL Number: MFCD00019722
Molecular Formula: C10H9BO2
Molecular Weight: 171.99
Form: Crystal
Color: White
Melting point (°C): 211


Other Product Names / Synonyms: 1-Naphthylboronic Acid

Formula: C₁₀H₉BO₂
Melting Pt: 211 °C
MDL Number: MFCD00019722
CAS Number: 13922-41-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bae4c25ee0bed755c653a0507bdc7b6ab891bbc&vid=8bae4c25ee0bed755c653a0507bdc7b6ab891bbc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR