2-Nitrocinnamaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 2'-Nitrocinnamaldehyde

N0611-25G N0611-5G
TCN0611-025GEA 370.19 CAD
TCN0611-025G TCN0611-5G
2-Nitrocinnamaldehyde ≥98.0%
2-Nitrocinnamaldehyde

CAS Number: 1466-88-2
MDL Number: MFCD00007188
Molecular Formula: C9H7NO3
Molecular Weight: 177.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 128


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 3-(2-Nitrophenyl)acrylaldehyde, 3-(2-Nitrophenyl)-2-propenal

Formula: C₉H₇NO₃
MW: 177.16 g/mol
Melting Pt: 128 °C
MDL Number: MFCD00007188
CAS Number: 1466-88-2
EINECS: 215-988-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b323526a099f8a5fb8093ac70072e26fe0728e0d&vid=b323526a099f8a5fb8093ac70072e26fe0728e0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR