1,4-Naphthalenedicarboxylic acid ≥95.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Naphthalene-1,4-dicarboxylic acid

N0606-25G N0606-100G
TCN0606-025GEA 64.78 CAD
TCN0606-025G TCN0606-100G
1,4-Naphthalenedicarboxylic acid ≥95.0% (by titrimetric analysis)
1,4-Naphthalenedicarboxylic acid

CAS Number: 605-70-9
MDL Number: MFCD00014312
Molecular Formula: C12H8O4
Molecular Weight: 216.19
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 320

Formula: C₁₂H₈O₄
MW: 216.19 g/mol
Melting Pt: 320 °C
MDL Number: MFCD00014312
CAS Number: 605-70-9
EINECS: 210-094-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ddb454e5f30b38b71a584d2a22b0b3bc433f2a0&vid=6ddb454e5f30b38b71a584d2a22b0b3bc433f2a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR