Ethyl-4-nitrocinnamate ≥98.0%

Supplier: TCI America

N0598-5G N0598-25G
TCN0598-005GEA 79 CAD
TCN0598-005G TCN0598-025G
Ethyl-4-nitrocinnamate ≥98.0%
Ethyl 4-nitrocinnamate

CAS Number: 953-26-4
MDL Number: MFCD00007380
Molecular Formula: C11H11NO4
Molecular Weight: 221.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 138


Other Product Names / Synonyms: 4-Nitrocinnamic Acid Ethyl Ester

Formula: C₁₁H₁₁NO₄
Melting Pt: 138 °C
MDL Number: MFCD00007380
CAS Number: 953-26-4
EINECS: 213-463-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38e1526c7c72fc03e6cc5b28c94cb040219cc504&vid=38e1526c7c72fc03e6cc5b28c94cb040219cc504&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR