cis-3-Nonen-1-ol ≥95.0%

Supplier: TCI America

N0588-10ML N0588-25ML
TCN0588-010MLEA 267.02 CAD
TCN0588-010ML TCN0588-25ML
cis-3-Nonen-1-ol ≥95.0%
cis-3-Nonen-1-ol

CAS Number: 10340-23-5
MDL Number: MFCD00010314
Molecular Formula: C9H18O
Molecular Weight: 142.24
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 109
Flash Point (°C): 102
Specific Gravity (20/20): 0.85

Formula: C₉H₁₈O
Boiling Pt: 109 °C
Density: 0.85 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00010314
CAS Number: 10340-23-5
EINECS: 233-735-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b60d550ded9b4ca5cca492f119519e4b5424613&vid=6b60d550ded9b4ca5cca492f119519e4b5424613&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR