5-Nitroisatin ≥98.0%

Supplier: TCI America

Synonyms: 5-Nitro-1H-indole-2,3-dione, 5-Nitroindoline-2,3-dione

N0572-5G N0572-25G
TCN0572-005GEA 60.04 CAD
TCN0572-005G TCN0572-25G
5-Nitroisatin ≥98.0%
5-Nitroisatin

CAS Number: 611-09-6
MDL Number: MFCD00005720
Molecular Formula: C8H4N2O4
Molecular Weight: 192.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 255


Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction)
Other Product Names / Synonyms: 5-Nitroindole-2,3-dione, 5-Nitro-2,3-indolinedione

Formula: C₈H₄N₂O₄
Melting Pt: 255 °C
MDL Number: MFCD00005720
CAS Number: 611-09-6
EINECS: 210-252-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=511a7dc3351a61ea3316386ad2c0af52ed2f3faa&vid=511a7dc3351a61ea3316386ad2c0af52ed2f3faa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR