Dimethyl-5-nitroisophthalate

Supplier: TCI America

Synonyms: Dimethyl 5-nitroisophthalate, 5-Nitroisophthalic acid dimethyl ester

N0567-500G N0567-25G
TCN0567-500GEA 299.25 CAD
TCN0567-500G TCN0567-025G
Dimethyl-5-nitroisophthalate
Dimethyl 5-nitroisophthalate

CAS Number: 13290-96-5
MDL Number: MFCD00008429
Molecular Formula: C10H9NO6
Molecular Weight: 239.18
Form: Crystal
Color: White
Melting point (°C): 124
Flash Point (°C): 110


Other Product Names / Synonyms: 5-Nitroisophthalic Acid Dimethyl Ester

Formula: C₁₀H₉NO₆
Melting Pt: 124 °C
Flash Pt: 110 °C
MDL Number: MFCD00008429
CAS Number: 13290-96-5
EINECS: 236-307-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af34769c1a38c6fd269803cc26c820ec1fe7d8d8&vid=af34769c1a38c6fd269803cc26c820ec1fe7d8d8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR