3-Nitrophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(3-Nitrophenyl)metaboric acid, Dihydroxy(3-nitrophenyl)borane, 3-Nitrobenzeneboronic acid
N0563-25G
N0563-5G
TCN0563-25GEA
314.1
CAD
TCN0563-25G
TCN0563-005G
3-Nitrophenylboronic acid (contains varying amounts of Anhydride)
3-Nitrophenylboronic acid
CAS Number: 13331-27-6
MDL Number: MFCD00007193
Molecular Formula: C6H6BNO4
Molecular Weight: 166.93
Form: Crystal
Color: White
Melting point (°C): 268
Other Product Names / Synonyms: 3-Nitrobenzeneboronic Acid
Formula:
C₆H₆BNO₄ MW: 166.93 g/mol Melting Pt: 268 °C |
MDL Number:
MFCD00007193 CAS Number: 13331-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...