4-Nitrobenzaldehyde ≥98.0%

Supplier: TCI America

N0559-25G N0559-500G N0559-100G
TCN0559-025GEA 57.39 CAD
TCN0559-025G TCN0559-500G TCN0559-100G
4-Nitrobenzaldehyde ≥98.0%
4-Nitrobenzaldehyde

CAS Number: 555-16-8
MDL Number: MFCD00007346
Molecular Formula: C7H5NO3
Molecular Weight: 151.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 105

Formula: C₇H₅NO₃
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 105 °C
Density: 1.546 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007346
CAS Number: 555-16-8
EINECS: 209-084-5
Merck Index: 12,06684
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=944429aa53135b1480f042068acf22b0bceabc16&vid=f2704327411add31565cb5c58ee388c99382237e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR