8-Nitroquinaldine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
N0539-5G
N0539-25G
TCN0539-005GEA
145.36
CAD
TCN0539-005G
TCN0539-025G
8-Nitroquinaldine ≥98.0% (by GC, titration analysis)
8-Nitroquinaldine
CAS Number: 881-07-2
MDL Number: MFCD00006764
Molecular Formula: C10H8N2O2
Molecular Weight: 188.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 138
Other Product Names / Synonyms: 8-Nitroquinaldine
Formula:
C₁₀H₈N₂O₂ Melting Pt: 138 °C |
MDL Number:
MFCD00006764 CAS Number: 881-07-2 EINECS: 212-919-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...