6-Nitroveratraldehyde ≥85.0%

Supplier: TCI America

Synonyms: 4,5-Dimethoxy-2-nitrobenzaldehyde, 3,4-Dimethoxy-6-nitrobenzaldehyde

N0538-5G N0538-25G
TCN0538-005GEA 178.54 CAD
TCN0538-005G TCN0538-25G
6-Nitroveratraldehyde ≥85.0%
6-Nitroveratraldehyde

CAS Number: 20357-25-9
MDL Number: MFCD00007134
Molecular Formula: C9H9NO5
Molecular Weight: 211.17
Purity/Analysis Method: >85.0% (GC)
Form: Crystal
Melting point (°C): 132


Product Categories: Bioscience, Inhibitors (for Pharmacological Research)
Other Product Names / Synonyms: 3,4-Dimethoxy-6-nitrobenzaldehyde

Formula: C₉H₉NO₅
MW: 211.17 g/mol
Melting Pt: 132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007134
CAS Number: 20357-25-9
EINECS: 243-762-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1361893d048039447e522f4fd2501d72ba9efd95&vid=1361893d048039447e522f4fd2501d72ba9efd95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR