4-Nitro-3-picoline-N-oxide ≥97.0%

Supplier: TCI America

Synonyms: 4-Nitropicoline N-oxide, 3-Methyl-4-nitropyridine 1-oxide, 3-Methyl-4-nitropyridine-N-oxide, 3-Methyl-4-nitropyridine N-oxide, 4-Nitro-3-picoline N-oxide, 3-Methyl-4-nitropyridine-1-oxide

N0527-5G N0527-25G
TCN0527-005GEA 183.44 CAD
TCN0527-005G TCN0527-25G
4-Nitro-3-picoline-N-oxide ≥97.0%
4-Nitro-3-picoline N-oxide

CAS Number: 1074-98-2
MDL Number: MFCD00014626
Molecular Formula: C6H6N2O3
Molecular Weight: 154.13
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 137


Product Categories: Materials Chemistry, Organic Nonlinear Optical (NLO) Materials
Other Product Names / Synonyms: 4-Nitro-3-picoline N-Oxide

Formula: C₆H₆N₂O₃
Melting Pt: 137 °C
MDL Number: MFCD00014626
CAS Number: 1074-98-2
EINECS: 214-050-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcfa1d2a34ad6354a5e7cfea673e5cabcf20f57f&vid=fcfa1d2a34ad6354a5e7cfea673e5cabcf20f57f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR