(±)-1-(1-Naphthyl)ethylamine ≥98.0%

Supplier: TCI America

Synonyms: 1-(Naphthalen-1-yl)ethanamine

N0523-500ML N0523-25ML
TCN0523-500MLEA 2286.26 CAD
TCN0523-500ML TCN0523-025ML
(±)-1-(1-Naphthyl)ethylamine ≥98.0%
(±)-1-(1-Naphthyl)ethylamine

CAS Number: 42882-31-5
MDL Number: MFCD00004014
Molecular Formula: C12H13N
Molecular Weight: 171.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 110
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: DL-alpha-(1-Aminoethyl)naphthalene

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 142 °C
Density: 1.07 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004014
CAS Number: 42882-31-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2cca6ceee8c92fce033733dd262c8aa516dfe5e&vid=c2cca6ceee8c92fce033733dd262c8aa516dfe5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR