5-Nitroisophthalic acid ≥99.0% (by HPLC)

Supplier: TCI America

N0520-25G N0520-500G
TCN0520-025GEA 33.18 CAD
TCN0520-025G TCN0520-500G
5-Nitroisophthalic acid ≥99.0% (by HPLC)
5-Nitroisophthalic acid

CAS Number: 618-88-2
MDL Number: MFCD00007254
Molecular Formula: C8H5NO6
Molecular Weight: 211.13
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 261
Flash Point (°C): 120

Formula: C₈H₅NO₆
MW: 211.13 g/mol
Melting Pt: 261 °C
Flash Pt: 120 °C
MDL Number: MFCD00007254
CAS Number: 618-88-2
EINECS: 210-568-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85225b77c38e56d4f6293e1f7d9d9b1c01fc4469&vid=85225b77c38e56d4f6293e1f7d9d9b1c01fc4469&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR