4-Nitroimidazole ≥98.0%

Supplier: TCI America
Warning

Synonyms: 4-Nitro-1H-imidazole, 5-Nitroimidazole, 5-Nitro-1H-imidazole

N0506-25G N0506-500G
TCN0506-025GEA 50.56 CAD
TCN0506-025G TCN0506-500G
4-Nitroimidazole ≥98.0%
4-Nitroimidazole

CAS Number: 3034-38-6
MDL Number: MFCD00005196
Molecular Formula: C3H3N3O2
Molecular Weight: 113.08
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 312
Flash Point (°C): 200

Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
Melting Pt: 312 °C
Flash Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00005196
CAS Number: 3034-38-6
EINECS: 221-224-7

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR