Dimethyl-2-nitroterephthalate ≥97.0%

Supplier: TCI America

N0501-250G N0501-25G
TCN0501-250GEA 197.5 CAD
TCN0501-250G TCN0501-025G
Dimethyl-2-nitroterephthalate ≥97.0%
Dimethyl 2-nitroterephthalate

CAS Number: 5292-45-5
MDL Number: MFCD00008422
Molecular Formula: C10H9NO6
Molecular Weight: 239.18
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 75
Flash Point (°C): 110


Other Product Names / Synonyms: Nitroterephthalic Acid Dimethyl Ester

Formula: C₁₀H₉NO₆
Melting Pt: 75 °C
Flash Pt: 110 °C
MDL Number: MFCD00008422
CAS Number: 5292-45-5
EINECS: 226-134-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ded79d7a083a728097b96daf1cadb36356bc776&vid=2ded79d7a083a728097b96daf1cadb36356bc776&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR