1-Iodoperfluorbutane ≥98.0%

Supplier: TCI America

N0499-25G N0499-500G N0499-100G
TCN0499-025GEA 67.5 CAD
TCN0499-025G TCN0499-500G TCN0499-100G
1-Iodoperfluorbutane ≥98.0%
1-Iodoperfluorbutane

CAS Number: 423-39-2
MDL Number: MFCD00001062
Molecular Formula: C4F9I
Molecular Weight: 345.93
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 67
Specific Gravity (20/20): 2.06


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Perfluorobutyl Iodide

Formula: C₄F₉I
Boiling Pt: 67 °C
Melting Pt: –2 °C
Density: 2.06 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001062
CAS Number: 423-39-2
EINECS: 207-025-8
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43953aab63e8baf3604a1d7ac50ddf8967ffbe1c&vid=fbb35dae343a0056fec64c77248ffeb441e372e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR