2-Methoxy-5-nitrophenol ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2-Hydroxy-4-nitroanisole5-Nitroguaiacol

N0498-500G N0498-25G
TCN0498-500GEA 654.59 CAD
TCN0498-500G TCN0498-025G
2-Methoxy-5-nitrophenol ≥98.0% (by titrimetric analysis)
2-Methoxy-5-nitrophenol

CAS Number: 636-93-1
MDL Number: MFCD00015561
Molecular Formula: C7H7NO4
Molecular Weight: 169.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 112
Melting point (°C): 105


Other Product Names / Synonyms: 2-Methoxy-5-nitrophenol, 2-Hydroxy-1-methoxy-4-nitrobenzene

Formula: C₇H₇NO₄
MW: 169.14 g/mol
Boiling Pt: 112 °C
Melting Pt: 105 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00015561
CAS Number: 636-93-1
EINECS: 211-269-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f0207413d401d255b4f81475abac0d597e0e919&vid=5f0207413d401d255b4f81475abac0d597e0e919&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR