2-Nitro-p-toluonitrile ≥98.0%

Supplier: TCI America

Synonyms: 4-Methyl-2-nitrobenzonitrile

N0494-5G N0494-1G
TCN0494-5GEA 668.34 CAD
TCN0494-5G TCN0494-001G
2-Nitro-p-toluonitrile ≥98.0%
2-Nitro-p-toluonitrile

CAS Number: 26830-95-5
MDL Number: MFCD00056107
Molecular Formula: C8H6N2O2
Molecular Weight: 162.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 99


Other Product Names / Synonyms: 4-Methyl-2-nitrobenzonitrile

Formula: C₈H₆N₂O₂
MW: 162.15
Melting Pt: 99 °C
MDL Number: MFCD00056107
CAS Number: 26830-95-5
EINECS: 248-019-5
UN: 3439
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=457c33b35706b60abda7352d2bb27dd88c1f310a&vid=457c33b35706b60abda7352d2bb27dd88c1f310a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR