(4-Nitrophenyl)-α-D-galactopyranoside ≥98.0%
Supplier: TCI America
Synonyms:
(2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl α-D-galactopyranoside
N0492-1G
N0492-200MG
TCN0492-001GEA
229.1
CAD
TCN0492-001G
TCN0492-200MG
(4-Nitrophenyl)-α-D-galactopyranoside ≥98.0%
(4-Nitrophenyl) α-D-galactopyranoside
[Substrate for alpha-D-Galactosidase]
CAS Number: 7493-95-0
MDL Number: MFCD00065050
Molecular Formula: C12H15NO8
Molecular Weight: 301.25
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 168
Specific rotation [a]20/D: 239 deg (C=1, H2O)
Storage Temperature: 0-10°C
Formula:
C₁₂H₁₅NO₈ MW: 301.26 g/mol Melting Pt: 168 °C |
MDL Number:
MFCD00065050 CAS Number: 7493-95-0 EINECS: 231-343-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...