(R)-(+)-1-(1-Naphthyl)ethylamine ≥99.0%

Supplier: TCI America

Synonyms: (R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (+)-1-(1-Naphthyl)ethylamine

N0482-5G N0482-1G
TCN0482-005GEA 192.82 CAD
TCN0482-005G TCN0482-001G
(R)-(+)-1-(1-Naphthyl)ethylamine ≥99.0%
(R)-(+)-1-(1-Naphthyl)ethylamine

CAS Number: 3886-70-2
MDL Number: MFCD00064114
Molecular Formula: C12H13N
Molecular Weight: 171.24
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 110
Specific Gravity (20/20): 1.07
Specific rotation [a]20/D: 80.5 deg (neat)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (R)-(+)-alpha-(1-Aminoethyl)naphthalene

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 80 °C
Density: 1.07 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00064114
CAS Number: 3886-70-2
EINECS: 223-425-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6356a7c71212cfde1ca9719f174931b63c0dfc38&vid=6356a7c71212cfde1ca9719f174931b63c0dfc38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR