(S)-(-)-1-(1-Naphthyl)ethylamine ≥99.0%

Supplier: TCI America

Synonyms: (S)-(-)-alpha-(1-Naphthyl)ethylamine

N0481-5G N0481-1G
TCN0481-005GEA 181.64 CAD
TCN0481-005G TCN0481-001G
(S)-(-)-1-(1-Naphthyl)ethylamine ≥99.0%
(S)-(-)-1-(1-Naphthyl)ethylamine

CAS Number: 10420-89-0
MDL Number: MFCD00064179
Molecular Formula: C12H13N
Molecular Weight: 171.24
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 110
Specific Gravity (20/20): 1.07
Specific rotation [a]20/D: -80.5 deg (neat)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (S)-(-)-alpha-(1-Aminoethyl)naphthalene

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 125 °C
Density: 1.07 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00064179
CAS Number: 10420-89-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3215375c61373dfba91ceb9ebe8dbdef703e834a&vid=3215375c61373dfba91ceb9ebe8dbdef703e834a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR