3-Nitro-1H-1,2,4-triazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

N0477-5G N0477-1G
TCN0477-005GEA 55.3 CAD
TCN0477-005G TCN0477-1G
3-Nitro-1H-1,2,4-triazole ≥98.0% (by HPLC, titration analysis)
3-Nitro-1H-1,2,4-triazole

[Coupling Agent]
CAS Number: 24807-55-4
MDL Number: MFCD00009749
Molecular Formula: C2H2N4O2
Molecular Weight: 114.06
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 214
Storage Temperature: 0-10°C

Formula: C₂H₂N₄O₂
Melting Pt: 214 °C
Density: 1.65 g/cm³ (20 °C)
MDL Number: MFCD00009749
CAS Number: 24807-55-4
EINECS: 246-468-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=499b05958f703309225250ae4789d59c8078111f&vid=d7278656c6cd8fd9993dd1e8c8b3e91c77d6752d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR