1-Naphthyl phosphate monosodium salt monohydrate ≥97.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1-Naphthylphosphoric Acid Monosodium Salt Monohydrate, Monosodium 1-Naphthyl Phosphate Monohydrate, Sodium 1-Naphthyl Hydrogen Phosphate Monohydrate, Phosphoric Acid 1-Naphthyl Ester Monosodium Salt Monohydrate
[Substrate for Phosphatase]
CAS Number: 81012-89-7
MDL Number: MFCD00041008
Molecular Formula: C10H9O4P
Molecular Weight: 246.13
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 191
Product Categories: Bioscience, Alkali Phosphatase Substrates
Other Product Names / Synonyms: 1-Naphthylphosphoric Acid Monosodium Salt, Phosphoric Acid 1-Naphthyl Ester Monosodium Salt, Sodium 1-Naphthyl Hydrogen Phosphate
Formula:
C₁₀H₈NaO₄P·1H₂O MW: 264.15 g/mol Melting Pt: 191 °C Storage Temperature: Freezer |
MDL Number:
MFCD00150615 CAS Number: 81012-89-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...