Polyethylene glycol 400
Supplier: TCI America
N0443-25G
N0443-500G
TCN0443-025GEA
56.23
CAD
TCN0443-025G
TCN0443-500G
Polyethylene glycol 400
Polyethylene glycol 400
CAS Number: 25322-68-3
MDL Number: MFCD01779601
Form: Viscous Clear Liquid
Color: Colorless
Flash Point (°C): 224
Specific Gravity (20/20): 1.13
Other Product Names / Synonyms: PEG 400
Formula:
H(OCH₂CH₂)nOH Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: 4…8 °C Density: 1.13 (20 °C) Flash Pt: 224 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...