(±)-Nipecotic acid ≥98.0%

Supplier: TCI America

Synonyms: H-DL-Nip-OH, (±)-3-Piperidinecarboxylic acid, Piperidine-3-carboxylic acid, Nipecotic acid

N0420-25G N0420-500G
TCN0420-025GEA 101.21 CAD
TCN0420-025G TCN0420-500G
(±)-Nipecotic acid ≥98.0%
(±)-Nipecotic acid

CAS Number: 498-95-3
MDL Number: MFCD00005992
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Bioscience, Neurotransmitter Agent Ingredients, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: Nipecotic Acid

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD00005992
CAS Number: 498-95-3
EINECS: 207-873-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ee23314bbc50308e53175bf2fea70a33d6e9cc9&vid=3ee23314bbc50308e53175bf2fea70a33d6e9cc9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR