2-Nitrothiophene ≥80.0% (contains 3-Nitrothiophene)

Supplier: TCI America

N0407-5G N0407-25G
TCN0407-005GEA 50.56 CAD
TCN0407-005G TCN0407-025G
2-Nitrothiophene ≥80.0% (contains 3-Nitrothiophene)
2-Nitrothiophene

CAS Number: 609-40-5
MDL Number: MFCD00005425
Molecular Formula: C4H3NO2S
Molecular Weight: 129.13
Purity/Analysis Method: >80.0% (GC)
Form: Crystal
Boiling point (°C): 224
Melting point (°C): 43
Flash Point (°C): 93

Formula: C₄H₃NO₂S
MW: 129.14 g/mol
Boiling Pt: 224
Melting Pt: 46.5
Density: 1.3644 g/cm³ (42.6 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00005425
CAS Number: 609-40-5
EINECS: 210-190-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2e9ae19fec304a187961ba0aa7c7213c27c11db&vid=e2e9ae19fec304a187961ba0aa7c7213c27c11db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR