6-Nitroindole ≥98.0%

Supplier: TCI America

N0402-1G N0402-5G
TCN0402-001GEA 60.04 CAD
TCN0402-001G TCN0402-5G
6-Nitroindole ≥98.0%
6-Nitroindole

CAS Number: 4769-96-4
MDL Number: MFCD00051497
Molecular Formula: C8H6N2O2
Molecular Weight: 162.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 143

Formula: C₈H₆N₂O₂
Melting Pt: 143 °C
MDL Number: MFCD00051497
CAS Number: 4769-96-4
EINECS: 225-311-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3939d19722383c8e79ce852635d20caef6bf29d&vid=a3939d19722383c8e79ce852635d20caef6bf29d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR